Potassium citrate monohydrate ≥99%, granular USP, FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
Management of Change (MOC) category = R
3068-01
3068-05
JT3068-1CS
901.17
USD
JT3068-1
JT3068-5
Potassium citrate monohydrate ≥99%, granular USP, FCC, J.T.Baker®
Potassium citrate monohydrate
Granular.
Formula:
HOC(COOK)(CH₂COOK)₂·H₂O MW: 324.41 g/mol Melting Pt: 230 Density: 1.98 g/cm³ (20 °C) |
MDL Number:
MFCD00150442 CAS Number: 6100-05-6 EINECS: 212-755-5 Merck Index: 13,07706 |
|
Specification Test Results
GMP Manufactured Product | |
Food GMP Manufactured Product | |
Meets U.S.P Requirements | |
Meets F.C.C. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Identification | Passes Test |
USP - Alkalinity | Passes Test |
USP - Loss on Drying | 3.0 - 6.0 % |
USP - Tartrate | Passes Test |
USP - Assay (C₆H₅K₃O₇) (dried basis) | 99.0 - 100.5 % |
FCC - Identification | Passes Test |
FCC - Alkalinity | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Loss on Drying | 3.0 - 6.0 % |
FCC - Assay (C₆H₅K₃O₇) (dried basis) | 99.0 - 100.5 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Metallic Residues: No metal catalysts or metal reagents, as defined by | |
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of | |
this material. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...