Potassium chlorate ≥99.0% (by KMnO₄ titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

3024-07
JT3024-7EA 2318.5 USD
JT3024-7
Potassium chlorate ≥99.0% (by KMnO₄ titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium chlorate
Formula: KClO₃
MW: 122.55 g/mol
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 356 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 13,07703

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KClO₃) (by KMnO₄ titrn) ≥99.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C, For Information Only
Bromate (BrO₃) ≤0.01 %
Calcium (Ca) ≤0.002 %
Chloride (Cl) ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
Sulfate (SO₄) Passes Test
Sodium (Na) ≤0.010 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR