Potassium bromate ≥99.8% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

2992-01
JT2992-1CS 803.44 USD
JT2992-1
Potassium bromate ≥99.8% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Potassium bromate

Lot analysis on label.

Formula: KBrO₃
MW: 167 g/mol
Melting Pt: 350 °C
Density: 3.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 12,07779

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KBrO₃) (by iodometry) ≥99.8 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.0 - 9.0
Bromide (Br) Passes Test
Sulfate (SO₄) ≤0.005 %
Iron (Fe) ≤0.002 %
Sodium (Na) ≤0.010 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Appearance (clear, colorless liquid)
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR