Water, Biotech Reagent for injection (WFI), J.T.Baker®

Supplier: Avantor Performance Materials

Synonyms: Dihydrogen oxide

Filtered through a 0.2 µm filter.

2910-89
JT2910-89EA 1606.5 USD
JT2910-89
Water, Biotech Reagent for injection (WFI), J.T.Baker®
Water

Avantor’s Water for Injection (WFI) quality water can help supplement existing media & buffer preparation processes by providing a reliable, high-quality alternative to in-house water production. Our water for injection quality water is manufactured under cGMP manufacturing guidelines using a multi-effect distillation process, ozone sparge and UV treatment. It is passed through a 0.2 micron filter prior to filling.


  • Manufactured in accordance with ICH-Q7 cGMP guidelines
  • Meets USP and EP specifications.
  • Packaged in bulk (200 L) for commercial use


Caution: For manufacturing, processing, or repackaging. Not intended for parenteral use.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Appearance (Clear & Colorless liquid) Passes Test
Bacterial Endotoxins (EU/mL) <=0.25 EU/mL
Bioburden <=10 CFU/mL
pH @ 25° C 5 to 7
Total Organic Carbon (TOC) recorded from recirculation loop <200 ppm
Nitrates (NO3) <0.2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR