Sulfuric acid 96%, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

2874-R
JT2874-REA 3.87 USD
JT2874-R MK2874-R
Sulfuric acid 96%, AR® ACS, Macron Fine Chemicals™
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (H₂SO₄) 95.0 - 98.0 %
ACS - Color (APHA) ≤8
Identification A Passes Test
Identification B Passes Test
ACS - Substances Reducing Permanganate (as SO₂) Passes Test
ACS - Residue after Ignition ≤4 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤0.5 ppm
ACS - Ammonium (NH₄) ≤1 ppm
ACS - Chloride (Cl) ≤0.1 ppm
ACS - Nitrate (NO₃) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤0.200 ppm
Trace Impurities - Arsenic (As) ≤0.004 ppm
Trace Impurities - Boron (B) ≤0.010 ppm
Trace Impurities - Chromium (Cr) ≤0.200 ppm
Trace Impurities - Copper (Cu) ≤0.10 ppm
Trace Impurities - Gold (Au) ≤0.300 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.300 ppm
Trace Impurities - Magnesium (Mg) ≤0.20 ppm
Trace Impurities - Manganese (Mn) ≤0.200 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Trace Impurities - Potassium (K) ≤0.300 ppm
Trace Impurities - Tin (Sn) ≤0.200 ppm
Trace Impurities - Titanium (Ti) ≤0.300 ppm
Trace Impurities - Zinc (Zn) ≤0.200 ppm
Trace Impurities - Mercury (Hg) ≤5.0 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR