Mineral oil, white stabilized USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Stabilization: Stabilised with vitamin E 10-100 ppm
Management of Change (MOC) category = R

2705-01
JT2705-1CS 1217.39 USD
JT2705-1
Mineral oil, white stabilized USP, J.T.Baker®
Mineral oil

White. Stabilized with vitamin E.

Boiling Pt: 260 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Acidity Passes Test
USP - Limit of Polycyclic Aromatic Hydrocarbons Passes Test
USP - Limit of Sulfur Compounds Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.845 - 0.905
USP - Viscosity at 40 C, mm₂/s 34.5 - 150.0
USP - Readily Carbonizable Substances Passes Test
USP - Solid Paraffin Passes Test
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR