Magnesium chloride hexahydrate ≥98%, crystals USP, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Magnesium dichloride
Management of Change (MOC) category = HR
2449-01
2449-07
2449-08
2449-R
JT2449-1CS
1607.61
USD
JT2449-1
JT2449-7
JT2449-08
JT2449-R
Magnesium chloride hexahydrate ≥98%, crystals USP, Multi-Compendial
Magnesium chloride hexahydrate
Crystals. GMP manufactured. Pyrogen tested.
Formula:
MgCl₂·6H₂O MW: 203.3 g/mol Boiling Pt: 1412 Melting Pt: 117 Density: 1.57 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 13,05684 |
Specification Test Results
Meets B.P. Chemical Specifications | |
GMP Manufactured Product | |
Meets E.P. Chemical Specifications | |
Meets U.S.P Requirements | |
Preserve in Tight Containers | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - pH (1 in 20) | 4.5 - 7.0 |
USP - Insoluble Matter | ≤ 0.005 % |
USP - Sulfate (SO₄) | ≤ 0.005 % |
USP - Barium (Ba) | Passes Test |
USP - Calcium (Ca) | ≤ 0.01 % |
USP - Potassium (K) | Passes Test |
USP - Assay | 98.0 - 101.0 % |
USP - Endotoxin Concentration (EU/g) | ≤ 2.5 |
EP/BP - Assay | 98.0 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Acidity or Alkalinity | Passes Test |
EP/BP - Bromide (Br) | ≤ 500 ppm |
EP/BP - Sulfate (SO₄) | ≤ 100 ppm |
EP/BP - Calcium (Ca) | ≤ 0.1 % |
EP/BP - Iron (Fe) | ≤ 10 ppm |
EP/BP - Water (H₂O) | 51.0 - 55.0 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...