Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

Management of Change (MOC) category = N

2425-07
JT2425-07CS 18718.46 USD
JT2425-07
Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Density: 1.45 g/cm³
MDL Number: MFCD00149214
CAS Number: 16674-78-5

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃COO)₂Mg·4H₂O) 98.0 - 102.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 6.5 - 8.5
ACS - Chloride (Cl) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Barium (Ba) ≤0.001 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Manganese (Mn) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Sodium (Na) ≤0.005 %
ACS - Strontium (Sr) ≤0.005 %
ACS - Endotoxin Concentration (2.5 EU/g max.) Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR