Lithium chloride ≥99.0% (by argentometric titration), granular, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
2370-01
2370-05
2370-07
JT2370-1CS
1215.84
USD
JT2370-1
JT2370-5
JT2370-7
Lithium chloride ≥99.0% (by argentometric titration), granular, BAKER ANALYZED® ACS
Lithium chloride
Granular. Lot analysis on label.
Formula:
LiCl Boiling Pt: 1382 °C (1013 hPa) Melting Pt: 614 °C Density: 2.07 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 EINECS: 231-212-3 Merck Index: 13,05550 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (LiCl) (by Ag titrn) | ≥99.0 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Loss on Drying | ≤1.0 % |
ACS - Titrable Base (meq/g) | ≤0.008 |
ACS - Sulfate (SO₄) | ≤0.01 % |
ACS - Barium (Ba) | ≤0.002 % |
ACS - Heavy Metals (as Pb) | ≤0.002 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - Calcium (Ca) | ≤0.01 % |
ACS - Potassium (K) | ≤0.01 % |
ACS - Sodium (Na) | ≤0.20 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...