Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

Management of Change (MOC) category = R

2262-20
JT2262-20EA 5106.18 USD
JT2262-20
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, ACS, J.T.Baker®
D(+)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00148912
CAS Number: 50-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C: Water (H₂O) ≤1.0 %
USP - Assay (Anhydrous basis) 97.5 - 102.0 %
USP - Related substances: Maltose and isomaltose ≤0.4 %
USP - Dextrin Passes Test
USP - Related substances: Maltotriose ≤0.2 %
USP - Related substances: Fructose ≤0.15 %
USP - Related substances: Unspecified impurities, each ≤0.10 %
USP - Related substances: Total Impurities ≤0.5 %
USP - Color and Clarity of Solution Passes Test
USP - Soluble Starch, Sulfites ≤15 ppm
USP - Conductivity (uS cm-1) ≤20 µS/cm
Chloride (Cl) ≤0.01 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Insoluble Matter ≤0.005 %
Loss on Drying at 105°C ≤0.2 %
Residue after Ignition ≤0.015 %
Solution in Water (50%)(APHA) ≤20
Specific Rotation [Å]²⁵ ^D (+) 52.5 - 53.0 Degree
Starch Passes Test
Sulfate and Sulfite (as SO₄) ≤0.005 %
Titrable Acid (meq/g) ≤0.002
Trace Impurities - Iron (Fe) ≤5 ppm
Appearance (white, granular powder)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR