Lanthanum(III) chloride heptahydrate 64.5 - 70.0% (by anhydrous basis), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = N
2255-05
JT2255-5CS
1294.58
USD
JT2255-5
JT2255-07
Lanthanum(III) chloride heptahydrate 64.5 - 70.0% (by anhydrous basis), BAKER ANALYZED® ACS, J.T.Baker®
Lanthanum (III) chloride heptahydrate
For flame enhancement in atomic absorption spectroscopy. Lot analysis on label.
For flame enhancement in atomic absorption spectroscopy.
Formula:
LaCl₃·7H₂O Boiling Pt: 860 °C (1013 hPa) Melting Pt: 857 °C Density: 3.8773 g/cm³ (–0.15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149756 CAS Number: 10025-84-0 EINECS: 233-237-5 Merck Index: 13,05379 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (LaCl₃) | 64.5 - 70.0 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Calcium (Ca) | ≤0.001 % |
Trace Impurities - Magnesium (Mg) | ≤10 ppm |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...