Hydrogen peroxide 30% stabilized, Electronic Grade, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

2204-01
JT2204-1CS 390.28 USD
JT2204-1
Hydrogen peroxide 30% stabilized, Electronic Grade, J.T.Baker®
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: <0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

For Microelectronic Use
Assay (H₂O₂) 30.0 - 32.0 %
Color (APHA) ≤10
Free Acid (µeq/g) ≤0.2
Residue after Evaporation ≤15 ppm
Ammonium (NH₄) ≤3 ppm
Chloride (Cl) ≤2 ppm
Nitrate (NO₃) ≤2 ppm
Phosphate (PO₄) ≤2 ppm
ACS - Sulfate (SO₄) ≤5 ppm
Trace Impurities - Aluminum (Al) ≤200.0 ppb
Trace Impurities - Antimony (Sb) ≤10.0 ppb
Trace Impurities - Arsenic (As) ≤10.0 ppb
Arsenic and Antimony (as As) ≤10.0 ppb
Trace Impurities - Barium (Ba) ≤20.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤20.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤50.0 ppb
Trace Impurities - Chromium (Cr) ≤20.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤20.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤10.0 ppb
Heavy Metals (as Pb) ≤500.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Niobium (Nb) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤600.0 ppb
Trace Impurities - Silicon (Si) ≤100.0 ppb
Trace Impurities - Silver (Ag) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤500.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤50.0 ppb
Trace Impurities - Tin (Sn) ≤1000.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤50 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR