Hydrogen peroxide solution 30% stabilized, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

2189-01
JT2189-1CS 2437.66 USD
JT2189-1
Hydrogen peroxide solution 30% stabilized, J.T.Baker®
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₂O₂) 29.0 - 32.0 %
Residue after Evaporation ≤0.0300 %
Nitrogen Compounds (as N) ≤0.001 %
Phosphate (PO₄) ≤0.02 %
Sulfate (SO₄) ≤0.001 %
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR