L(+)-Glutamic acid 98.5-101.5% USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

Management of Change (MOC) category = R

2077-06
JT2077-6EA 277.83 USD
JT2077-6
L(+)-Glutamic acid 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Assay (C₅H₉NO₄) (dried basis) 98.5 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 30.5 - 32.5 Degree
EP/BP - Ninhydrin-Positive Substances Passes Test
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤200 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
USP - Assay (C₅H₉NO₄) (dried basis) 98.5 - 101.5 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.02 %
USP - Sulfate (SO₄) ≤0.02 %
USP - Iron (Fe) ≤10 µg/g
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
USP - Specific Rotation [Å]²º^D (+) 31.5 - 32.5 Degree
USP - Loss on Drying at 105°C ≤0.1 %
USP - Identification Passes Test
Endotoxin Concentration, EU/g ≤3.0
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR