Ammonium iron (II) sulfate hexahydrate 98.5-101.5% (by KMnO₄ titration), fine crystals, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

Management of Change (MOC) category = N

2054-05 2054-01
JT2054-5CS 6064.18 USD
JT2054-5 JT2054-1 JT2054-7 35001-222 JT2054-69
Ammonium iron (II) sulfate hexahydrate 98.5-101.5% (by KMnO₄ titration), fine crystals, BAKER ANANYZED™ ACS
Ammonium iron (II) sulphate hexahydrate

Fine crystals. Lot analysis on label.


Caution: Due to inherent oxidation, ferric ion (Fe³⁺) content may increase on storage.

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: 100 °C
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Fe(NH₄)₂(SO₄)₂·6H₂O) (by KMnO₄ titrn) 98.5 - 101.5 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 3.0 - 5.0
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.003 %
Ferric Ion (Fe³⁺) ≤0.01 %
ACS - Magnesium (Mg) ≤0.002 %
Manganese (Mn) ≤0.01 %
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
NOTE: Due to inherent oxidation, ferric ion (Fe³⁺) content may increase
on storage.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR