L-Arginine hydrochloride, Multi-Compendial, J.T.Baker®

Supplier: Avantor Performance Materials

2051-06 2051-07 2051-09
JT2051-6EA 548.01 USD
JT2051-6 JT2051-7 JT2051-9
L-Arginine hydrochloride, Multi-Compendial, J.T.Baker®
L-Arginine hydrochloride

US sourced amino acids


Caution: Storage Condition - Preserve in well closed containers. Store in well-closed, light resistant containers.

Formula: C₆H₁₅ClN₄O₂
Melting Pt: 218 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Order Now

Specification Test Results

USP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.5 %
USP - Identification Passes test
USP - Specific Rotation [Å]²º^D (+) 21.4 - 23.6 °
USP - Loss on Drying at 105°C ≤ 0.2 %
USP - Residue on Ignition ≤ 0.1 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
USP - Chloride (Cl) 16.5 - 17.1 %
EP/BP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes test
EP/BP - Identification B Passes test
EP/BP - Identification E Passes test
EP/BP - Appearance of Solution Passes test
EP/BP - Specific Rotation [Å]²º^D (+) 21.0 - 23.5 °
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C₆H₁₄N₄O₂·HCl) (dried basis) 98.5 - 101.0 %
JP - Identification Passes test
JP - Optical Rotation (+) 21.5 - 23.5 °
JP - pH (1 in 10) 4.7 - 6.2
JP - Clarity and Color of Solution Passes test
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Related Substances Passes test
JP - Loss on Drying at 105°C ≤ 0.20 %
JP - Residue after Ignition ≤ 0.10 %
Endotoxin Concentration, IU/mg For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR