Iron(III) nitrate nonahydrate 98.0-101.0% (by iodometry), BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

Management of Change (MOC) category = N

2018-01 2018-05
JT2018-1CS 733.48 USD
JT2018-1 JT2018-5
Iron(III) nitrate nonahydrate 98.0-101.0% (by iodometry), BAKER ANALYZED® ACS
Iron (III) nitrate nonahydrate

Crystals. Lot analysis on label.

Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125
Melting Pt: 47
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 13,04057

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Fe(NO₃)₃·9H₂O) (by iodometry) 98.0 - 101.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤5 ppm
Sulfate (SO₄) ≤0.01 %
Calcium (Ca) ≤0.01 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.05 %
Manganese (Mn) ≤0.01 %
pH of 5% Solution at 25°C 1.5 - 2.5
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR