Ammonium iron (III) citrate (14.5 - 16.0% Fe), green, powder FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = RL

1977-01
JT1977-1CS 834.39 USD
JT1977-1
Ammonium iron (III) citrate (14.5 - 16.0% Fe), green, powder FCC, J.T.Baker®
Ammonium iron (III) citrate

Green powder.

Formula: C₆H₅+4y)Fe(x)N(y)O₇
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6
Merck Index: 13,00518

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Assay (as Fe) 14.5 - 16.0 %
FCC - Identification Passes Test
FCC - Ferric Citrate Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg) ≤1 mg/kg
FCC - Oxalate (C₂O₄) Passes Test
FCC - Sulfate (SO₄) ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR