Dextrose, anhydrous, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

Management of Change (MOC) category = N

1916-07 1916-QM
JT1916-7EA 1086.85 USD
JT1916-7 JT1916-QM
Dextrose, anhydrous, powder, BAKER ANALYZED® ACS, J.T.Baker®
D(+)-Glucose

Powder. Lot analysis on label.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Specific Rotation [Å]²⁵ ^D (+) 52.5 - 53.0 Degree
Insoluble Matter ≤0.005 %
Loss on Drying at 105°C ≤0.2 %
Residue after Ignition ≤0.015 %
Solution in Water (50%)(APHA) ≤20
Titrable Acid (meq/g) ≤0.002
Chloride (Cl) ≤0.01 %
Sulfate and Sulfite (as SO₄) ≤0.005 %
Starch Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR