Calcium chloride dihydrate 99.0-105.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

1332-01 1332-07 1332-05
JT1332-1CS 588.6 USD
JT1332-1 JT1332-7 JT1332-5 JT1332-69
Calcium chloride dihydrate 99.0-105.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CaCl₂) 74.0 - 78.0 %
Assay (CaCl₂·2H₂O) 99.0 - 105.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 4.5 - 8.5
Oxidizing Substances (as NO₃) ≤0.003 %
Sulfate (SO₄) ≤0.01 %
Ammonium (NH₄) ≤0.005 %
Barium (Ba) ≤0.005 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.1 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR