Calcium chloride ≥89%, pellets, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

Management of Change (MOC) category = N

1313-05 1313-01 1313-07
JT1313-5CS 1883.55 USD
JT1313-5 JT1313-1 JT1313-7
Calcium chloride ≥89%, pellets, purified, J.T.Baker®
Calcium chloride

Pellets. Desiccant, 4 to 8 mesh.

Formula: CaCl₂
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.15 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CaCl₂) ≥89 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.02 %
Trace Impurities - Arsenic (As) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR