Diboron trioxide ≥98.0%, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Boron oxide, Boric anhydride

Management of Change (MOC) category = N

1176-05
JT1176-5CS 2849.34 USD
JT1176-5
Diboron trioxide ≥98.0%, purified, J.T.Baker®
di-Boron trioxide

Powder.

Formula: B₂O₃
Boiling Pt: 1860 °C (1013 hPa)
Melting Pt: 450 °C
Density: 1.81…1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (B₂O₃) (ignited basis) ≥98.0 %
Loss on Ignition ≤10.0 %
Sulfate (SO₄) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR