Barium sulfate 97.5-100.5% USP, Multi-Compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

1041-01
JT1041-1CS 1720.06 USD
JT1041-1
Barium sulfate 97.5-100.5% USP, Multi-Compendial
Barium sulphate

Fine, white powder.

Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600
Melting Pt: 1345
Density: 4.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Bulkiness Passes Test
USP - pH of 10% Aqueous Solution at 25°C 3.5 - 10.0
USP - Sulfide (µg/g) ≤0.5
USP - Acid-Soluble Substances ≤0.3 %
USP - Soluble Barium Salts ≤0.001 %
USP - Assay 97.5 - 100.5 %
USP - Particulate Matter Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Acid-Soluble Substances ≤0.3 %
EP/BP - Oxidizable Sulfur Compounds Passes Test
EP/BP - Soluble Barium Salts ≤10 ppm
EP/BP - Phosphate (PO₄) ≤50 ppm
EP/BP - Arsenic (As) ≤2 ppm
EP/BP - Loss on Ignition ≤2.0 %
EP/BP - Sediment Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR