L(+)-Ascorbic acid 99.0-100.5%, fine-granular USP, FCC 20 - 80 Mesh, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
Management of Change (MOC) category = R
0938-08
0938-05
0938-07
JT0938-8CS
4640.61
USD
JT0938-8
JT0938-5
JT0938-7
L(+)-Ascorbic acid 99.0-100.5%, fine-granular USP, FCC 20 - 80 Mesh, J.T.Baker®
L(+)-Ascorbic acid
Fine granules. 20 to 80 mesh.
- Food GMP manufactured product
Formula:
C₆H₈O₆ MW: 176.13 g/mol Boiling Pt: 553 °C (1013 hPa) Melting Pt: 190…192 °C Density: 1.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 EINECS: 200-066-2 REACH: exempt Merck Index: 13,00837 |
Specification Test Results
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Meets U.S.P Requirements | |
Food GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
USP - Assay (C₆H₈O₆) | 99.0 - 100.5 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Specific Rotation [Å]²⁵ ^D (+) | 20.5 - 21.5 Degree |
USP - Residue on Ignition | ≤0.1 % |
FCC - Assay (C₆H₈O₆) | 99.0 - 100.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Specific Rotation [Å]²⁵ ^D (+) | 20.5 - 21.5 Degree |
FCC - Residue on Ignition | ≤0.1 % |
Arsenic (As) | ≤3 ppm |
Mesh - Thru U.S. No. 20 Sieve | ≥100 % |
Mesh - Thru U.S. No. 30 Sieve | ≥95 % |
Mesh - Thru U.S. No. 80 Sieve | ≤20 % |
Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3 | |
Solvents are likely to be present. Residual Class 2 Solvents are below | |
Option 1 and the residual concentration of all Class 3 Solvents is | |
<0.5%. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...