Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = R
0897-07
0897-09
0897-79
JT0897-7EA
3176.86
USD
JT0897-7
JT0897-9
JT0897-79
Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
Store above 16°C (60°F) to prevent freezing | |
CAUTION: For Manufacturing, processing or repackaging | |
Assay (NaOH) | 50 - 52 % |
Chloride (Cl) | ≤30 ppm |
Heavy Metals (as Pb) | ≤1 ppm |
Iron (Fe) | ≤2 ppm |
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications | |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Filtered through a 5 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...