Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = R

0895-07 0895-08
JT0895-7EA 2180.63 USD
JT0895-7 JT0895-08 JT0895-QM
Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Water for injection quality water.

Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: 1.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
Protect from air to avoid absorption of carbon dioxide.
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 4.9 - 5.1
Chloride (Cl) ≤10 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Iron (Fe) ≤2 ppm
Endotoxin Concentration (EU/mL ≤2
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR