Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = R
0895-07
0895-08
JT0895-7EA
2180.63
USD
JT0895-7
JT0895-08
JT0895-QM
Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Water for injection quality water.
Formula:
NaOH Boiling Pt: 109 °C (1013 hPa) Density: 1.18 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
Protect from air to avoid absorption of carbon dioxide. | |
CAUTION: For Manufacturing, processing or repackaging | |
Appearance (Clear, colorless liquid) | Passes Test |
Normality | 4.9 - 5.1 |
Chloride (Cl) | ≤10 ppm |
ACS - Heavy Metals (as Pb) | ≤1 ppm |
Iron (Fe) | ≤2 ppm |
Endotoxin Concentration (EU/mL | ≤2 |
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications | |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Filtered through a 0.2 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...