Ammonia solution 14%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = RL

0889-15
JT0889-15CS 4515.43 USD
JT0889-15
Ammonia solution 14%, J.T.Baker®
Ammonia solution
Formula: NH₃
Density: 0.90 g/cm³
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (as NH₃) 11.2 - 16.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR