Ammonium hydrogen difluoride ≥96.0%, soluble, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

0619-07
JT0619-7EA 1152.76 USD
JT0619-7 JT0619-9
Ammonium hydrogen difluoride ≥96.0%, soluble, BAKER ANALYZED®
Ammonium hydrogen difluoride
Formula: NH₄HF₂
Melting Pt: 124.6 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NH₄F·HF) ≥96.0 %
Insoluble Matter ≤0.02 %
pH of 5% Solution at 25°C 4.0 - 6.5
Heavy Metals (as Pb) ≤0.002 %
Moisture (as H₂O) ≤0.4 %
Product may contain black specks that are indigenous to product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR