Ammonium acetate ≥97% for HPLC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = NI
0599-08
JT0599-8CS
1491.59
USD
JT0599-8
Ammonium acetate ≥97% for HPLC, J.T.Baker®
Ammonium acetate
For high performance liquid chromatography. Lot analysis on label.
Formula:
H₃CCOONH₄ Melting Pt: 114 °C Density: 1.17 g/cm³ (20 °C) Flash Pt: 136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 EINECS: 211-162-9 Merck Index: 13,00495 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃COONH₄) (by acid-base titration) | ≥97 % |
Insoluble Matter | ≤0.005 % |
Residue after Ignition | ≤0.010 % |
pH of 5% Solution at 25°C | 6.7 - 7.3 |
Nitrate (NO₃) | ≤0.001 % |
Sulfate (SO₄) | ≤0.001 % |
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 350 nm | ≤0.010 |
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm | ≤0.010 |
UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm | ≤0.020 |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...