Glycine 98.5-101.5% USP, FCC

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

Management of Change (MOC) category = R

0581-01 0581-05 0581-07
JT0581-1CS 824.32 USD
JT0581-1 JT0581-5 JT0581-7
Glycine 98.5-101.5% USP, FCC
Glycine
Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets F.C.C. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Loss on Drying ≤0.2 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.007 %
USP - Sulfate (SO₄) ≤0.0065 %
USP - Hydrolyzable Substances Passes Test
USP - Related Compounds: Glycine anhydride ≤0.1 %
USP - Related Compounds: Monochloroacetic acid (For Information Only)
USP - Related Compounds: Iminodiacetic acid ≤0.1 %
USP - Related Compounds: Diglycine ≤0.1 %
USP - Related Compounds: Triglycine ≤0.1 %
USP - Related Compounds: Hexamethylenetetramine ≤0.1 %
USP - Related Compounds: Any unspecified impurity (For Information Only)
USP - Related Compounds: Total impurities ≤1.0 %
FCC - Assay (C₂H₅NO₂) (dried basis) 98.5 - 101.5 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤5 mg/kg
FCC - Loss on Drying ≤0.2 %
FCC - Residue on Ignition ≤0.1 %
FCC - Insoluble Foreign Matter Passes Test
Only Class 2 Solvents (Methanol) are likely to be present. All are
below Option 1 limits.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR