Glycine 98.5-101.5% USP, FCC
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
Management of Change (MOC) category = R
0581-01
0581-05
0581-07
JT0581-1CS
824.32
USD
JT0581-1
JT0581-5
JT0581-7
Glycine 98.5-101.5% USP, FCC
Glycine
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: 83 °C Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Specification Test Results
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Food GMP Manufactured Product | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemical | |
USP - Assay (C₂H₅NO₂) (dried basis) | 98.5 - 101.5 % |
USP - Identification | Passes Test |
USP - Loss on Drying | ≤0.2 % |
USP - Residue on Ignition | ≤0.1 % |
USP - Chloride (Cl) | ≤0.007 % |
USP - Sulfate (SO₄) | ≤0.0065 % |
USP - Hydrolyzable Substances | Passes Test |
USP - Related Compounds: Glycine anhydride | ≤0.1 % |
USP - Related Compounds: Monochloroacetic acid (For Information Only) | |
USP - Related Compounds: Iminodiacetic acid | ≤0.1 % |
USP - Related Compounds: Diglycine | ≤0.1 % |
USP - Related Compounds: Triglycine | ≤0.1 % |
USP - Related Compounds: Hexamethylenetetramine | ≤0.1 % |
USP - Related Compounds: Any unspecified impurity (For Information Only) | |
USP - Related Compounds: Total impurities | ≤1.0 % |
FCC - Assay (C₂H₅NO₂) (dried basis) | 98.5 - 101.5 % |
FCC - Identification | Passes Test |
FCC - Lead (Pb) | ≤5 mg/kg |
FCC - Loss on Drying | ≤0.2 % |
FCC - Residue on Ignition | ≤0.1 % |
FCC - Insoluble Foreign Matter | Passes Test |
Only Class 2 Solvents (Methanol) are likely to be present. All are | |
below Option 1 limits. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...