Aluminium oxide, powder, BAKER ANALYZED® for chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

Management of Change (MOC) category = N

0537-05 0537-01
JT0537-5CS 3159.28 USD
JT0537-5 JT0537-1
Aluminium oxide, powder, BAKER ANALYZED® for chromatography, J.T.Baker®
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
pH of 5% Slurry at 20°C 6.0 - 7.5
Adsorption, mg o-nitroaniline/g ≥0.05
Loss on Drying ≤5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR