Aluminium oxide ≥98.5%, powder, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

Management of Change (MOC) category = N

0536-05
JT0536-5CS 1272.76 USD
JT0536-5
Aluminium oxide ≥98.5%, powder, BAKER ANALYZED®, J.T.Baker®
Aluminium oxide

Neutral powder.

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Al₂O₃) ≥98.5 %
Loss on Ignition ≤1.0 %
Chloride (Cl) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR