Aluminium nitrate nonahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Aluminum nitrate nonahydrate

Management of Change (MOC) category = NI

0528-05
JT0528-5CS 2378.62 USD
JT0528-5
Aluminium nitrate nonahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS
Aluminium nitrate nonahydrate

Crystals. Lot analysis on label.


  • Lot analysis on label

Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1 III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Al(NO₃)₃·9H₂O) (by EDTA titrn) 98.0 - 102.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 2.5 - 3.5
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Magnesium (Mg) ≤0.001 %
ACS - Potassium (K) ≤0.002 %
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR