Sulfanilic acid, anhydrous 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Management of Change (MOC) category = N

0354-01
JT0354-1CS 7411.74 USD
JT0354-1
Sulfanilic acid, anhydrous 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Sulphanilic acid

For nitrogen dioxide determination (ASTM D-1607).


  • Lot analysis on label

Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 279 °C
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (4-NH₂C₆H₄SO₃H) 98.0 - 102.0 %
Residue after Ignition ≤0.010 %
Insoluble in Na₂CO₃ Solution ≤0.01 %
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Suitability for Nitrogen Dioxide Determination (ASTM D-1607) Passes Test
Nitrite (NO₂) ≤0.5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR