Stearic acid triple pressed ≥90.0%, powder NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Octadecanoic acid, Octadecylic acid

Management of Change (MOC) category = R

0340-R 0340-07 0340-01 0340-05
JT0340-REA 1737.72 USD
JT0340-R JT0340-7 JT0340-1 JT0340-5
Stearic acid triple pressed ≥90.0%, powder NF, J.T.Baker®
Stearic acid

Triple pressed to remove impurities.

Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 53…59 °C (as freezing point)
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B (Acid Value) 194 - 212
NF - Identification C Passes Test
NF - Residue on Ignition ≤0.1 %
NF - Acidity Passes Test
NF - Color of Solution Passes Test
NF - Iodine Value ≤4.0
NF - Freezing Point 53 - 59 °C
NF - Assay (C₁₈H₃₆O₂) (stearic acid) 40.0 - 60.0 %
NF - Assay (Total of Stearic & Palmitic) ≥90.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR