Sodium hydroxide 50% in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = R

0339-09 0339-07 BAKR0339-03 0339-26 0339-79 0339-69
JT0339-9EA 37849.45 USD
JT0339-9 JT0339-7 JT0339-3 MK0339-15 JT0339-26 JT0339-79 JT0339-69
Sodium hydroxide 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Biotech Reagent—A grade of product which is produced under GMP guidelines, but for which there is no USP/NF compendia for the product.


  • Filtered through a 5 micron filter


Made from USP purified water and sodium hydroxide pellets, NF (Multicompendial) which meets BP, Ph.Eur. and JP chemical specifications.

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
Store above 16°C (60°F) to prevent freezing
CAUTION: For Manufacturing, processing or repackaging
Assay (NaOH) 50 - 52 %
Chloride (Cl) ≤30 ppm
Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤2 ppm
Sodium Carbonate (Na₂CO₃) ≤3.0 %
Made from USP Purified Water and Sodium Hydroxide pellets, NF (Multicompendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 5 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR