Sodium hydroxide 25% in aqueous solution, BAKER ANANYZED™, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = R
0338-07
BAKR0338-09
0338-26
JT0338-7EA
924.79
USD
JT0338-7
JT0338-9
JT0338-26
Sodium hydroxide 25% in aqueous solution, BAKER ANANYZED™, Biotech Reagent, J.T.Baker®
Sodium hydroxide
Made from USP purified water and multicompendial sodium hydroxide pellets. Filtered through a 0.2 µm element. GMP manufactured product.
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Assay (NaOH) | 24.5 - 25.5 % |
Chloride (Cl) | ≤20 ppm |
Heavy Metals (as Pb) | ≤1 ppm |
Iron (Fe) | ≤2 ppm |
Endotoxin Concentration (EU/mL) | ≤2.0 |
Made from USP Purified Water and Sodium Hydroxide pellets, NF (Multicompendial) which meets BP, Ph.Eur. and JP Chemical Specifications | |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Filtered through a 0.2 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...