Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = R

0329-09 0329-07 0329-26
JT0329-9EA 7208.15 USD
JT0329-9 JT0329-7 JT0329-26
Sodium hydroxide 0.5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Made from USP purified water and sodium hydroxide pellets, NF (Multi-Compendial) which meets BP, Ph. Eur. and JP chemical specifications.

Formula: NaOH
Boiling Pt: 105 °C (1013 hPa)
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
Protect from air to avoid absorption of carbon dioxide.
CAUTION: For Manufacturing, processing or repackaging
Appearance (clear, colorless solution) Passes Test
Normality 0.480 - 0.520
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Endotoxin Concentration (EU/mL) ≤2.0
Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR