Sodium hydroxide 1.0 N in aqueous solution, BAKER ANANYZED™, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Filtered through a 0.2 µm filter.

0328-09 0328-07 0328-08
JT0328-9EA 7270.78 USD
JT0328-9 JT0328-7 JT0328-08
Sodium hydroxide 1.0 N in aqueous solution, BAKER ANANYZED™, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.

Formula: NaOH
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (clear, colorless solution) Passes Test
Normality 0.975 - 1.025
Chloride (Cl) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.5 ppm
Endotoxin Concentration (EU/mL) ≤ 2.0
Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR