Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = N
0324-05
0324-01
JT0324-5EA
1736.12
USD
JT0324-5
JT0324-1
Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®
Silicic acid hydrate
Powder.
Formula:
SiO₂·xH₂O MW: 60.09 g/mol (anhy) Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1710 °C Density: 2.10 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00054122 CAS Number: 1343-98-2 REACH: 01-2119379499-16 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (as SiO₂) | ≥84 % |
Nonvolatile with HF | ≤0.4 % |
Chloride (Cl) | ≤0.01 % |
Sulfate (SO₄) | ≤0.02 % |
Heavy Metals (as Pb) | ≤0.002 % |
Iron (Fe) | ≤0.007 % |
Loss on Ignition (as H₂O) | ≤16 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...