Hydrochloric acid 25%, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = R

0323-07
JT0323-07CS 1341.37 USD
JT0323-07
Hydrochloric acid 25%, Biotech Reagent, J.T.Baker®
Hydrochloric acid

Made from USP purified water and hydrochloric acid, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.


Filtered through a 0.2 micron filter.

Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless) Passes Test
Assay (HCl) 24.0 - 26.0 %
Residue after Evaporation ≤0.0100 %
Arsenic (As) ≤1.500 ppm
Sulfate (SO₄) ≤1 ppm
Ammonium (NH₄) ≤3 ppm
Heavy Metals (as Pb) ≤0.5 ppm
Iron (Fe) ≤0.1 ppm
Made from U.S.P. Purified Water and Hydrochloric Acid, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR