Orthophosphoric acid ≥85-88% (by acidimetry) NF, ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Phosphoric acid

Management of Change (MOC) category = R

0240-07
JT0240-7EA 8036.45 USD
JT0240-7
Orthophosphoric acid ≥85-88% (by acidimetry) NF, ACS
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Meets N.F. Requirements
GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (H₃PO₄) 85.0 - 88.0 %
NF - Identification Passes Test
NF - Nitrate (NO₃) Passes Test
NF - Phosphorous or Hypophosphorous Acid Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Alkali Phosphates Passes Test
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Calcium (Ca) ≤0.002 %
Chloride (Cl) ≤3 ppm
Color (APHA) ≤10
Heavy Metals (as Pb) ≤8 ppm
Insoluble Matter ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
Nitrate (NO₃) ≤5 ppm
Reducing Substances Passes Test
Sulfate (SO₄) ≤12 ppm
Trace Impurities - Antimony (Sb) ≤20.000 ppm
Trace Impurities - Arsenic (As) ≤0.500 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Trace Impurities - Manganese (Mn) ≤0.500 ppm
Trace Impurities - Potassium (K) ≤40.000 ppm
Trace Impurities - Sodium (Na) ≤200.000 ppm
Volatile Acids (as CH₃COOH) ≤0.001 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR