Lactic acid 88.0-92.0%, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

0200-01 0200-03 0200-07 0200-09
JT0200-01CS 2312.94 USD
JT0200-01 JT0200-03 JT0200-07 JT0200-09
Lactic acid 88.0-92.0%, Multi-Compendial, J.T.Baker®
Lactic acid
Formula: H₃CCH(OH)COOH
Density: ∼1.20 g/ml (20 °C)
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (CH₃CHOHCOOH) 88.0 - 92.0 %
USP - Chloride (Cl) Passes Test
USP - Citric, Oxalic, Phosphoric, or Tartaric Acids Passes Test
USP - Identification Passes Test
USP - Readily Carbonizable Substances Passes Test
USP - Residue on Ignition ≤ 0.05 %
USP - Sugars Passes Test
USP - Sulfate (SO₄) Passes Test
EP - Assay (CH₃CHOHCOOH) 88.0 - 92.0 %
EP - Appearance Passes Test
EP - (S)-enantiomer ≥ 95.0 %
EP - Calcium (Ca) ≤ 200 ppm
EP - Citric, Oxalic, and Phosphoric Acids Passes Test
EP - Endotoxin Concentration, <5 IU/g Passes Test
EP - Ether-Insoluble Substances Passes Test
EP - Identification A Passes Test
EP - Identification B Passes Test
EP - Identification C Passes Test
EP - Methanol (by GC) ≤ 50 ppm
EP - Sugars and Other Reducing Substances Passes Test
EP - Sulfate (SO₄) ≤ 200 ppm
EP - Sulfated Ash ≤ 0.1 %
JP - Assay (CH₃CHOHCOOH) 85.0 - 92.0 %
JP - Chloride (Cl) ≤ 0.036 %
JP - Citric, Oxalic, Phosphoric, or Tartaric Acids Passes Test
JP - Cyanide (CN) Passes Test
JP - Glycerin and Mannitol Passes Test
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Identification Passes Test
JP - Iron (Fe) ≤ 5 ppm
JP - Readily Carbonizable Substances Passes Test
JP - Residue on Ignition ≤ 0.10 %
JP - Sugars Passes Test
JP - Sulfate (SO₄) ≤ 0.010 %
JP - Volatile Fatty Acids Passes Test
JP - Optical Rotation -52 - -46 Degree
This product is pyrogen free.
Only Class 3 Solvents are likely to be present. Residual concentration
of all Class 3 Solvents is <0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR