Citric acid monohydrate 99.5-100.5% (by anhydrous basis), crystalline powder FCC, USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

Management of Change (MOC) category = R

0120-05
JT0120-5CS 2669.82 USD
JT0120-5
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), crystalline powder FCC, USP, J.T.Baker®
Citric acid monohydrate

Powder.

Formula: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 135…152 °C
Density: 1.54 g/cm³
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification (USP) Passes Test
USP - Water (H₂O) (U.S.P.) 7.5 - 9.0 %
USP - Residue on Ignition ≤0.05 %
USP - Oxalic Acid (USP) ≤0.036 %
USP - Sulfate (SO₄) ≤0.015 %
USP - Readily Carbonizable Substances Passes Test
FCC - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification (FCC) Passes Test
FCC - Lead (Pb) ≤0.5 mg/kg
FCC - Oxalate (C₂O₄) (FCC) ≤100 mg/kg
FCC - Readily Carbonizable Substances Passes Test
FCC - Residue on Ignition ≤0.05 %
FCC - Water (H₂O) (F.C.C.) ≤8.8 %
Must be subjected to further processing during the preparation of
injectable dosage forms.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR