Orthoboric acid, granular NF, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Boron trihydroxide, Orthoboric acid, Boron hydroxide, Boric acid, Boracic acid
Management of Change (MOC) category = R
0096-05
JT0096-05CS
2429.58
USD
JT0096-05
Orthoboric acid, granular NF, J.T.Baker®
ortho-Boric acid
Formula:
B(OH)₃ MW: 61.83 g/mol |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 |
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Completeness of Solution | Passes Test |
NF - Identification | Passes Test |
NF - Solubility in Alcohol | Passes Test |
NF - Loss on Drying | ≤0.5 % |
NF - Assay (H₃BO₃) (dried basis) | 99.5 - 100.5 % |
Iron (Fe) | ≤4.5 ppm |
Total Transition Metals (Cr, Co, Cu, Mn) | ≤10 ppm |
Not for Internal Use | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...