I-CHEM™ High-Density Polyethylene Bottles and Jars, with Caps, Thermo Scientific
Supplier: Thermo Scientific
311-0500 311-1000 312-0250 312-0500 312-0950 313-0250 313-0500 313-1000 319-0125 319-1000 312-2000 312-0120BPC 312-0250BPC 312-0500BPC 312-0950BPC 312-4000 312-4000BPC 313-0125
IR311-0500CS 237.19 USD
IR311-0500 IR311-1000 IR312-0250 IR312-0500 IR312-0950 IR313-0250 IR313-0500 IR313-1000 IR319-0125 IR319-1000 IR-312-2000 IR113-0125BPC IR113-0500BPC IR113-1000BPC IR312-0120BPC IR312-0250BPC IR312-0500BPC IR312-0950BPC IR312-2000BPC IR312-4000 IR312-4000BPC IR313-0125
I-CHEM™ High-Density Polyethylene Bottles and Jars, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
HDPE containers, white, with LDPE-lined polypropylene cap, narrow/wide neck. Suitable for metals analyses and water quality parameters.
  • Narrow mouth for easy handling and precision pouring
  • Wide mouth for easy filling and sample retrieval
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Each container individually barcoded with lot number and unique container number
I-CHEM Certified™ 300 series containers are processed to meet or exceed 'USEPA Specifications and Guidance for Contaminent-Free Containers'. These containers are recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes. Certified to meet EPA performance based specifications for metals, cyanide, fluoride and other water quality parameters. Or unprocessed and readily assembled for own cleaning procedure.
Ordering information: Supplied with polyfoam-lined caps. Also available unprocessed and bulk packed for added economy.
Delivery information: Certificate of analysis (CofA) included in the case and barcoded with lot number for traceability. Analysis labels included unattached for optional use.
Packaging: The Carton is sealed with security tape providing tamper evidence. Each container is individually barcoded with lot number and unique container number.