Antibody Diluent, Normal, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
08980641
IC980641EA 116.45 USD
IC980641
Antibody Diluent, Normal, MP Biomedicals
Buffers Antibody Purification Buffers
Ready-to-use diluent for primary antibodies and secondary antibodies
Antibodies diluted in this reagent can be stored between 4-8°C for up to 18 months.