DNA-Erase™ Decontamination Solution, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
DNA-Erase™ is used to decontaminate laboratory surfaces, laboratory apparatus, plastic and glass vessels, and pipettors.
- Non-alkaline
- Non-corrosive
- Non-carcinogenic
- Decontamination reagent
- Ready to use formula
DNA-Erase™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against DNA contamination and is intended for use at PCR workstations for eliminating PCR artifacts from all laboratory surfaces, including benchtops, apparatus and pipettors. Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. The solution contains a surfactant plus an agent that destroys DNA. DNA-Erase™ is stable and heat resistant. DNA-Erase™ is ready-to-use for eliminating DNA from any surface. By following the simple decontamination instructions, DNA is completely destroyed and removed.
Ready to use
DNA-Erase™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against DNA contamination and is intended for use at PCR workstations for eliminating PCR artifacts from all laboratory surfaces, including benchtops, apparatus and pipettors .Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. The solution contains a surfactant plus an agent that destroys DNA. DNA-Erase™ is stable and heat resistant.DNA-Erase™ is ready-to-use for eliminating DNA from any surface. By following the simple decontamination instructions below, DNA is completely destroyed and removed.